Learn More
Sino Biological Recombinant Human REG4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11186H08S100
Encompass_Preferred
A DNA sequence encoding the human REG4 (NP_001152824.1) (Met 1-Pro 158) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6639