Learn More
Sino Biological Recombinant Human CDKL2 Protein (His Tag)
Supplier:  Sino Biological 11191H07B50
Encompass_Preferred
A DNA sequence encoding the human CDKL2 (Q92772) (Met 1-His 493) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6653