Learn More
Sino Biological Recombinant Human CSRP1 Protein (His Tag)
Supplier:  Sino Biological 11201H08E50
Encompass_Preferred
A DNA sequence encoding the human CSRP1 (NP_004069.1) (Met 1-Glu 193) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-161-6663