Learn More
Sino Biological Recombinant Human CD96 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11202H08H100
Encompass_Preferred
A DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met 1-Met 503) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6664