Learn More
Sino Biological Recombinant Human SETD7 Protein (His Tag)
Supplier:  Sino Biological 11209H07E100
Encompass_Preferred
A DNA sequence encoding the human SETD7 (NP_085151.1) (Asp 2-Lys 366) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6670