Learn More
Sino Biological Influenza A H7N7 (A/chicken/Netherlands/1/03) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11212V08H1100
Encompass_Preferred
A DNA sequence encoding an N-terminal segment (Met 1-Arg 344) of Influenza A virus (A/chicken/Netherlands/1/03 (H7N7)) hemagglutinin (AAR02639.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6678