Learn More
Sino Biological Recombinant Human CADM3 Protein (His Tag)
Supplier:  Sino Biological 11214H08H100
Encompass_Preferred
A DNA sequence encoding the human CADM3 isoform 2 (short isoform) (NP_001120645.1) extracellular domain (Met 1-His 330) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6684