Learn More
Sino Biological Recombinant Human RIZ1 Protein (GST Tag)
Supplier:  Sino Biological 11216H09E100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment of human PRDM2 (NP_036363.2) (Met 1-Ala 200), containing the SET domian, was fused with the GST tag at the N-terminus.
Catalog No. 50-161-6688