Learn More
Sino Biological Recombinant Human RIZ1 Protein
Supplier:  Sino Biological 11216HNCE100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment of human PRDM2 (NP_036363.2) (Met 1-Ala 200), containing the SET domian, was expressed, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-6690