Learn More
Sino Biological Recombinant Human SMYD3 Protein (GST Tag)
Supplier:  Sino Biological 11217H09B50
Encompass_Preferred
A DNA sequence encoding the human SMYD3 isoform 2 (NP_073580.1) (Lys 35-Ser 369) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-6693