Learn More
Sino Biological Recombinant Human Apolipoprotein H Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11221H08H100
Encompass_Preferred
A DNA sequence encoding the human APOH (NP_000033.2) (Met 1-Cys 345) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6702