Learn More
Sino Biological Recombinant Human AGO1 Protein (His Tag)
Supplier:  Sino Biological 11225H07B50
Encompass_Preferred
A DNA sequence encoding the full length of human AGO1 (NP_036331.1) (Met 1-Ala 857) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6713