Learn More
Sino Biological Recombinant Human OSMR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11226H08H100
Encompass_Preferred
A DNA sequence encoding the human OSMR (NP_003990.1) extracellular domain (Met 1-Met 740) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6714