Learn More
Sino Biological Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11229V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/1073/99(H9N2)) hemagglutinin (NP_859037.1) (Met1-Arg335), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6722