Learn More
Sino Biological Recombinant Human, Mouse HIST1H3A Protein
Supplier:  Sino Biological 11231HNAE100
Encompass_Preferred
A DNA sequence encoding the native human / mouse HIST1H3A (NP_003520.1) (Met1-Ala136) was expressed. Human and Mouse HIST1H3A sequences are identical.
Catalog No. 50-161-6727