Learn More
Sino Biological Recombinant Human FOLR1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11241H08H100
Encompass_Preferred
A DNA sequence encoding the human FOLR1 (NP_057937.1) (Met 1-Met 233) without the pro peptide was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6740