Learn More
Sino Biological Recombinant Human PPM1G Protein (aa 317-546, His Tag)
Supplier:  Sino Biological 11245H07E100
Encompass_Preferred
A DNA sequence encoding the human PPM1G (O15355) C-terminal fragment (Met 317-Asp 546) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6744