Learn More
Sino Biological Recombinant Human TGF-alpha Protein, HPLC-verified
Supplier:  Sino Biological 11252HNAE1
Encompass_Preferred
A DNA sequence encoding the human TGFa (P01135-1) (Val40-Ala89) was expressed and purified.
Catalog No. 50-161-6748