Learn More
Sino Biological Recombinant Human Peroxiredoxin 2 Protein (His Tag)
Supplier:  Sino Biological 11255H07B100
Encompass_Preferred
A DNA sequence encoding the human PRDX2 (P32119) (Met 1-Asn 198) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6752