Learn More
Sino Biological Recombinant Human Peroxiredoxin 6 Protein (His Tag)
Supplier:  Sino Biological 11257H07E100
Encompass_Preferred
A DNA sequence encoding the human PRDX6 (P30041) (Met 1-Pro 224) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6754