Learn More
Sino Biological Recombinant Human ACOX1 Protein (His Tag)
Supplier:  Sino Biological 11266H07B100
Encompass_Preferred
A DNA sequence encoding the human ACOX1 (AAH08767.1) (Met 1-Leu 660) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6760