Learn More
Sino Biological Recombinant Human CD63 Protein (His Tag)
Supplier:  Sino Biological 11271H08H100
Encompass_Preferred
A DNA sequence encoding the second extracellular domain of human CD63 (P08962-1) (Ala 103-Val 203) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-6762