Learn More
Sino Biological Recombinant Human G6B Protein (hFc Tag)
Supplier:  Sino Biological 11273H02H100
Encompass_Preferred
A DNA sequence encoding the human G6B (NP_612116.1) (Met1-Gln146) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6764