Learn More
Sino Biological Recombinant Human GPA33/A33 Protein (His Tag)
Supplier:  Sino Biological 11277H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human GPA33 (Q99795-1) (Met 1-Val 235) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6771