Learn More
Sino Biological Recombinant Human Adenylosuccinate Lyase Protein (His Tag)
Supplier:  Sino Biological 11287H07E100
Encompass_Preferred
A DNA sequence encoding the human ADSL (P30566-1) (Met 1-Leu 484) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6778