Learn More
Sino Biological Recombinant Human AKR1B1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11294H07E100
Encompass_Preferred
A DNA sequence encoding the human AKR1B1 (P15121) (Met 1-Phe 316) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6786