Learn More
Sino Biological Recombinant Human FLRT2 Protein (His Tag)
Supplier:  Sino Biological 11296H08H100
Encompass_Preferred
A DNA sequence encoding the human FLRT2 (O43155) extracellular domain (Met 1-Ser 539) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6788