Learn More
Sino Biological Recombinant Human Semaphorin 5A Protein (His Tag)
Supplier:  Sino Biological 11300H08H50
Encompass_Preferred
A DNA sequence encoding the human SEMA5A (NP_003957.2) extracellular domain (Met 1-Met 968) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6795