Learn More
Sino Biological Recombinant Human EED Protein (His & GST Tag)
Supplier:  Sino Biological 11307H20B100
Encompass_Preferred
A DNA sequence encoding the human EED (O75530-1) (Met 1-Arg 441) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-6800