Learn More
Sino Biological Recombinant Human EphA4 Protein (His & hFc Tag)
Supplier:  Sino Biological 11314H03H100
Encompass_Preferred
A DNA sequence encoding the human EPHA4 (NP_004429.1) extracellular domain (Met 1-Thr 547) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6810