Learn More
Sino Biological Recombinant Human FUT10 Protein (His Tag)
Supplier:  Sino Biological 11327H07H100
Encompass_Preferred
A DNA sequence encoding the lumenal domain of human FUT10 (Q6P4F1-1) (Leu 32-Asp 479) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6824