Learn More
Sino Biological Recombinant Human HSC70 Protein (His Tag)
Supplier:  Sino Biological 11329H07E100
Encompass_Preferred
A DNA sequence encoding the human HSPA8 (P11142-1) (Met 1-Asp 646) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-6826