Learn More
Sino Biological Recombinant Human Cpn10/HSPE1 Protein (His Tag)
Supplier:  Sino Biological 11330H07E50
Encompass_Preferred
A DNA sequence encoding the human HSPE1 (NP_002148.1) (Met1-Asp102) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6829