Learn More
Sino Biological Recombinant Human ORP150 Protein (His Tag)
Supplier:  Sino Biological 11342H08H50
Encompass_Preferred
A DNA sequence encoding the C-terminal segment of human HSP12A (NP_001124463.1) (Met 695-Leu 994) was expressed, fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-6835