Learn More
Sino Biological Recombinant Human Cadherin-17 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11360H08H100
Encompass_Preferred
A DNA sequence encoding the human CDH17 (Q12864-1) extracellular domain (Met 1-Met 787) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6836