Learn More
Sino Biological Recombinant Human BIN1 Protein (His Tag)
Supplier:  Sino Biological 11361H07E100
Encompass_Preferred
A DNA sequence encoding the human BIN1 isoform BIN1-13 (O00499-10) (Met 1-Pro 424) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6838