Learn More
eMolecules​ 1137-42-4 | 4-Hydroxybenzophenone | Ambeed | MFCD00002355 | 198.221 | C13H10O2 | 98.000 | Oc1ccc(cc1)C(=O)c1ccccc1 | 25g | 552550144
Catalog No. NC3926859 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Hydroxybenzophenone | Ambeed | 1137-42-4 | MFCD00002355 | 198.221 | C13H10O2 | 98.000 | Oc1ccc(cc1)C(=O)c1ccccc1 | 25g | 552550144
Catalog No. NC3926859 Supplier eMolecules​ Supplier No. 552550144
4-Hydroxybenzophenone | Ambeed | 1137-42-4 | MFCD00002355 | 198.221 | C13H10O2 | 98.000 | Oc1ccc(cc1)C(=O)c1ccccc1 | 25g | 552550144