Learn More
Sino Biological Recombinant Human MGAT5 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11373H08H50
Encompass_Preferred
A DNA sequence encoding the human MGAT5 (NP_002401.1) C-terminal segment (Leu 189-Leu 741) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the terminus.
Catalog No. 50-161-6849