Learn More
Sino Biological Recombinant Human LRRTM4 Protein (His Tag)
Supplier:  Sino Biological 11387H08H100
Encompass_Preferred
A DNA sequence encoding the human LRRTM4 extracellular domain (NP_001128217.1) (Met 1-Lys 424) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6853