Learn More
Sino Biological Recombinant Human ALDH1A1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11388H07E50
Encompass_Preferred
A DNA sequence encoding the human ALDH1A1 (NP_000680.2) (Ser 2-Ser 501) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6856