Learn More
Sino Biological Recombinant Human FLRT1 Protein (His Tag)
Supplier:  Sino Biological 11389H08H100
Encompass_Preferred
A DNA sequence encoding the human FLRT1 extracellular domain (Q9NZU1-1) (Met 1-Pro 524) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6857