Learn More
Sino Biological Recombinant Human Galanin Protein (hFc Tag)
Supplier:  Sino Biological 11392H02H100
Encompass_Preferred
A DNA sequence encoding the human GAL (P22466) (Met1-Ser123) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6859