Learn More
Sino Biological Recombinant Human CHST11 Protein (His Tag)
Supplier:  Sino Biological 11396H08B100
Encompass_Preferred
A DNA sequence encoding the human CHST11 (Q9NPF2-2) (Met36-Glu347) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6861