Learn More
Sino Biological Recombinant Human SULT2B1 Protein (His Tag)
Supplier:  Sino Biological 11410H07E50
Encompass_Preferred
A DNA sequence encoding the human SULT2B1 (NP_814444.1) (Asp 2-Ser 365) was expressed, with a N-terminal polyhistidine tag.
Catalog No. 50-161-6872