Learn More
Sino Biological Recombinant Human B3GNT2 Protein (hFc Tag)
Supplier:  Sino Biological 11416H01H50
Encompass_Preferred
A DNA sequence encoding the human B3GNT2 (Q9NY97-1) (Lys29-Cys397) was expressed,with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-6878