Learn More
Sino Biological Recombinant Human Pyruvate kinase PKM Protein (Isoform M2, His Tag)
Supplier:  Sino Biological 11430H07E50
Encompass_Preferred
A DNA sequence encoding the human PKM2 isoform M2 (P14618-1) (Ser 2-Pro 531) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-6883