Learn More
Sino Biological Recombinant Human STIM1 Protein (His Tag)
Supplier:  Sino Biological 11434H08H100
Encompass_Preferred
A DNA sequence encoding the human STIM1 (NP_003147.2) extracellular domain (Met 1-Asp 213) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6889