Learn More
Sino Biological Recombinant Human EphA3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11459H08H100
Encompass_Preferred
A DNA sequence encoding the human EPHA3 (NP_005224.2) (Met1-Gln541) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6897