Learn More
Sino Biological Recombinant Human CUTC Protein (His Tag)
Supplier:  Sino Biological 11470H07E100
Encompass_Preferred
A DNA sequence encoding the human CUTC (Q9NTM9) (Met 1-Val 273) was expressed, with an N-terminal polyhistidine tag.
Catalog No. 50-161-6901