Learn More
Sino Biological Recombinant Human IL-21 Receptor/IL-21R Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11483H08H100
Encompass_Preferred
A DNA sequence encoding the human IL21R (Q9HBE5-1) extracellular domain (Met 1-Pro 236) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6907